Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EDR2
Entrez Gene ID
827642
Gene Name
ENHANCED DISEASE RESISTANCE 2
Gene Aliases
AT4G19040, ENHANCED DISEASE RESISTANCE 2, F13C5.210, F13C5_210
Chromosome Location
Chr.4: 10431515 - 10437564 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203840.1 NP_001190769.1 6 - 7 769 87
NM_001341322.1 NP_001319993.1 6 - 7 902 87
NM_118022.5 NP_193639.2 6 - 7 902 87

View Full Product Details