Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- INDH
- Entrez Gene ID
-
827696
- Gene Name
- INDH1(iron-sulfur protein required for NADH dehydrogenase)
- Gene Aliases
-
AT4G19540, F24J7.100, F24J7_100, IND1(iron-sulfur protein required for NADH dehydrogenase)-like, INDL, iron-sulfur protein required for NADH dehydrogenase
- Chromosome Location
-
Chr.4: 10657289 - 10659566 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118074.2 |
NP_193689.1 |
1 - 2 |
302 |
96 |
GenBank mRNA |
AK229020.1 |
- |
1 - 2 |
132 |
96 |
View Full Product Details