Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLAH2
Entrez Gene ID
828910
Gene Name
SLAC1 homologue 2
Gene Aliases
AT4G27970, SLAC1 homologue 2, T13J8.80, T13J8_80
Chromosome Location
Chr.4: 13918165 - 13920378 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341891.1 NP_001320078.1 3 - 4 1202 80
NM_001341893.1 NP_001328004.1 2 - 3 1288 80
NM_001341895.1 NP_001328006.1 1 - 2 1488 80
NM_118935.3 NP_567792.1 2 - 3 1402 80
GenBank mRNA AF367337.1 - 2 - 3 1156 80
BT002238.1 - 2 - 3 1146 80

View Full Product Details