Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PIP5K1
- Entrez Gene ID
-
838801
- Gene Name
- phosphatidylinositol-4-phosphate 5-kinase 1
- Gene Aliases
-
AT1G21980, ATPIP5K1, ATPIPK1, PHOSPHATIDYLINOSITOL-4-PHOSPHATE 5-KINASE, phosphatidylinositol-4-phosphate 5-kinase 1
- Chromosome Location
-
Chr.1: 7734661 - 7738772 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102047.4 |
NP_173617.1 |
1 - 2 |
1490 |
118 |
GenBank mRNA |
AK221279.1 |
- |
1 - 2 |
1412 |
118 |
View Full Product Details