Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHR42
Entrez Gene ID
832164
Gene Name
chromatin remodeling 42
Gene Aliases
AT5G20420, F7C8.10, F7C8_10, chromatin remodeling 42
Chromosome Location
Chr.5: 6898790 - 6904242 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122049.3 NP_197542.1 3 - 4 484 111

View Full Product Details