Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RIN1
Entrez Gene ID
832293
Gene Name
P-loop containing nucleoside triphosphate hydrolases superfamily protein
Gene Aliases
AT5G22330, ATTIP49A, MWD9.12, MWD9_12, RESISTANCE TO PSEUDOMONAS SYRINGAE PV MACULICOLA INTERACTOR 1
Chromosome Location
Chr.5: 7390796 - 7394280 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122138.3 NP_197625.1 7 - 8 1124 76
GenBank mRNA AY084606.1 - 7 - 8 1038 76
BT000923.1 - 7 - 8 915 76

View Full Product Details