Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAM1
Entrez Gene ID
832365
Gene Name
methylthioalkylmalate synthase 1
Gene Aliases
2-ISOPROPYLMALATE SYNTHASE, 2-ISOPROPYLMALATE SYNTHASE 3, AT5G23010, IMS3, METHYLTHIOALKYLMALATE SYNTHASE, T20O7.3, T20O7_3, methylthioalkylmalate synthase 1
Chromosome Location
Chr.5: 7703041 - 7706973 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343784.1 NP_001330047.1 3 - 4 1049 65
NM_001343785.1 NP_001330048.1 3 - 4 998 65
NM_122207.3 NP_197692.1 3 - 4 998 65
GenBank mRNA AJ486884.1 - 3 - 4 917 65
AJ486885.1 - 3 - 4 917 65
AJ486886.1 - 3 - 4 917 65
AJ486887.1 - 3 - 4 917 65
AY049037.1 - 3 - 4 917 65
AY054203.1 - 3 - 4 989 65
AY070471.1 - 3 - 4 995 65

View Full Product Details