Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IMS2
Entrez Gene ID
832366
Gene Name
2-isopropylmalate synthase 2
Gene Aliases
2-isopropylmalate synthase 2, AT5G23020, MAM-L, MAM3, METHYLTHIOALKYMALATE SYNTHASE-LIKE, MYJ24.1, MYJ24_1
Chromosome Location
Chr.5: 7718118 - 7721866 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122208.4 NP_197693.1 8 - 9 1400 81
GenBank mRNA AJ486937.1 - 8 - 9 1315 81
AJ486942.1 - 8 - 9 1315 81
AJ486945.1 - 8 - 9 1315 81
AJ486946.1 - 8 - 9 1315 81
AY099549.1 - 8 - 9 1396 81
BT008874.1 - 8 - 9 1315 81

View Full Product Details