Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STP11
Entrez Gene ID
832391
Gene Name
sugar transporter 11
Gene Aliases
AT5G23270, ATSTP11, MKD15.13, MKD15_13, SUGAR TRANSPORTER 11, sugar transporter 11
Chromosome Location
Chr.5: 7838863 - 7841077 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122233.4 NP_197718.1 1 - 2 406 83
GenBank mRNA AJ344339.1 - 1 - 2 137 83
AK175890.1 - 1 - 2 362 83

View Full Product Details