Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FSD3
Entrez Gene ID
832395
Gene Name
Fe superoxide dismutase 3
Gene Aliases
AT5G23310, Fe superoxide dismutase 3, MKD15.17, MKD15_17, SOD3, superoxide dismutase 3
Chromosome Location
Chr.5: 7850444 - 7852534 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122237.4 NP_197722.1 7 - 8 853 82
GenBank mRNA AY065458.1 - 7 - 8 749 82
AY091225.1 - 7 - 8 673 82

View Full Product Details