Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDPK9
Entrez Gene ID
832424
Gene Name
calmodulin-like domain protein kinase 9
Gene Aliases
ARABIDOPSIS THALIANA CALMODULIN-LIKE DOMAIN PROTEIN KINASE 9, AT5G23580, ATCDPK9, ATCPK12, CALCIUM-DEPENDENT PROTEIN KINASE, CALCIUM-DEPENDENT PROTEIN KINASE 12, CPK12, calmodulin-like domain protein kinase 9
Chromosome Location
Chr.5: 7950202 - 7952619 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122264.5 NP_197748.1 3 - 4 1017 139
GenBank mRNA AK229078.1 - 3 - 4 890 139
BT026382.1 - 3 - 4 831 139
U20388.1 - 3 - 4 868 139

View Full Product Details