Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SQP2
Entrez Gene ID
832479
Gene Name
squalene monooxygenase 2
Gene Aliases
AT5G24140, MLE8.6, MLE8_6, squalene monooxygenase 2
Chromosome Location
Chr.5: 8164319 - 8167776 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122319.4 NP_197802.2 2 - 3 416 71

View Full Product Details