Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATXR6
Entrez Gene ID
832503
Gene Name
TRITHORAX-RELATED PROTEIN 6
Gene Aliases
ARABIDOPSIS TRITHORAX-RELATED PROTEIN 6, AT5G24330, MOP9.7, MOP9_7, SDG34, SET DOMAIN PROTEIN 34
Chromosome Location
Chr.5: 8295110 - 8297159 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122341.3 NP_197821.1 2 - 3 505 92
GenBank mRNA BT012223.1 - 2 - 3 409 92
BT012421.1 - 2 - 3 343 92

View Full Product Details