Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYCA2;1
Entrez Gene ID
832610
Gene Name
cyclin a2;1
Gene Aliases
AT5G25380, F18G18.15, F18G18_15, cyclin a2;1
Chromosome Location
Chr.5: 8814517 - 8817887 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343923.1 NP_001318646.1 9 - 10 1179 86
NM_001343924.1 NP_001332474.1 9 - 10 1179 86
NM_001343925.1 NP_001332473.1 8 - 9 981 86
NM_122447.3 NP_197920.2 8 - 9 963 86

View Full Product Details