Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AL4
Entrez Gene ID
832690
Gene Name
alfin-like 4
Gene Aliases
AT5G26210, T19G15.60, T19G15_60, alfin-like 4
Chromosome Location
Chr.5: 9158318 - 9160493 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122522.5 NP_197993.1 3 - 4 625 94
GenBank mRNA AB493760.1 - 3 - 4 353 94
AY087173.1 - 3 - 4 486 94
BT024475.1 - 3 - 4 353 94

View Full Product Details