Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PWD
Entrez Gene ID
832706
Gene Name
chloroplastidic phosphoglucan, water dikinase (ATGWD3)
Gene Aliases
AT5G26570, ATGWD3, F9D12.1, F9D12_1, OK1, PHOSPHOGLUCAN WATER DIKINASE
Chromosome Location
Chr.5: 9261479 - 9267732 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125808.1 NP_001119280.1 11 - 12 1929 130
NM_122538.5 NP_198009.3 11 - 12 2030 130
GenBank mRNA AJ635427.1 - 11 - 12 1929 130

View Full Product Details