Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IREG3
Entrez Gene ID
832740
Gene Name
iron-regulated protein 3
Gene Aliases
AT5G26820, ATIREG3, F2P16.6, F2P16_6, IRON REGULATED 3, MAR1, MULTIPLE ANTIBIOTIC RESISTANCE 1, RTS3, iron-regulated protein 3
Chromosome Location
Chr.5: 9433569 - 9437158 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122564.5 NP_198034.2 7 - 8 1312 64
GenBank mRNA AY062606.1 - 7 - 8 1264 64
AY128787.1 - 7 - 8 1023 64

View Full Product Details