Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GI
Entrez Gene ID
838883
Gene Name
gigantea protein (GI)
Gene Aliases
AT1G22770, FB, GIGANTEA, T22J18.6, T22J18_6
Chromosome Location
Chr.1: 8061751 - 8067790 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102124.3 NP_564180.1 13 - 14 3646 66
GenBank mRNA AF105064.1 - 12 - 13 3418 66
AJ133786.1 - 12 - 13 3208 66

View Full Product Details