Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LPR1
Entrez Gene ID
838909
Gene Name
Cupredoxin superfamily protein
Gene Aliases
AT1G23010, F19G10.5, F19G10_5, Low Phosphate Root1
Chromosome Location
Chr.1: 8147118 - 8149891 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102149.4 NP_173714.2 1 - 2 466 62

View Full Product Details