Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCA3
Entrez Gene ID
838983
Gene Name
beta carbonic anhydrase 3
Gene Aliases
AT1G23730, ATBCA3, BETA CARBONIC ANHYDRASE 3, BETA CARBONIC ANHYDRASE 3, F5O8.28, F5O8_28, beta carbonic anhydrase 3
Chromosome Location
Chr.1: 8395466 - 8398387 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332620.1 NP_001321762.1 7 - 8 1111 73
NM_102221.5 NP_173785.1 8 - 9 749 73
GenBank mRNA AY081265.1 - 8 - 9 682 73
AY114551.1 - 7 - 8 612 73

View Full Product Details