Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALDH2B7
Entrez Gene ID
838991
Gene Name
aldehyde dehydrogenase 2B7
Gene Aliases
ALDH2B, AT1G23800, F5O8.35, F5O8_35, aldehyde dehydrogenase 2B, aldehyde dehydrogenase 2B7
Chromosome Location
Chr.1: 8412051 - 8414886 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332622.1 NP_001321290.1 5 - 6 817 83
NM_102228.4 NP_564204.1 5 - 6 817 83
GenBank mRNA AY035139.1 - 5 - 6 809 83
AY113912.1 - 5 - 6 735 83

View Full Product Details