Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OVA9
Entrez Gene ID
839120
Gene Name
glutamine-tRNA ligase, putative / glutaminyl-tRNA synthetase, putative / GlnRS
Gene Aliases
AT1G25350, F4F7.26, F4F7_26, ovule abortion 9
Chromosome Location
Chr.1: 8889018 - 8894306 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198165.1 NP_001185094.1 7 - 8 676 100
NM_102345.4 NP_173906.2 7 - 8 721 100
GenBank mRNA AY062594.1 - 7 - 8 675 100
AY114653.1 - 7 - 8 620 100

View Full Product Details