Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RCN1
Entrez Gene ID
839135
Gene Name
ARM repeat superfamily protein
Gene Aliases
AT1G25490, ATB BETA BETA, EER1, ENHANCED ETHYLENE RESPONSE 1, F2J7.19, F2J7_19, PHOSPHOPROTEIN PHOSPHATASE 2A REGULATORY SUBUNIT A, REGA, ROOTS CURL IN NPA, SERINE/THREONINE PROTEIN PHOSPHATASE TYPE 2A REGULATORY SUBUNIT A
Chromosome Location
Chr.1: 8951162 - 8955160 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102360.4 NP_173920.1 4 - 5 933 101
GenBank mRNA AY120757.1 - 4 - 5 875 101
BT000108.1 - 3 - 4 758 101
U21557.1 - 4 - 5 836 101

View Full Product Details