Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HK3
Entrez Gene ID
839621
Gene Name
histidine kinase 3
Gene Aliases
AHK3, AT1G27320, F17L21.11, F17L21_11, histidine kinase 3
Chromosome Location
Chr.1: 9486889 - 9492632 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102494.4 NP_564276.1 3 - 4 1851 70
GenBank mRNA AB046870.1 - 3 - 4 960 70

View Full Product Details