Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATCSA-1
Entrez Gene ID
839677
Gene Name
Transducin/WD40 repeat-like superfamily protein
Gene Aliases
AT1G27840, F28L5.15, F28L5_15
Chromosome Location
Chr.1: 9693008 - 9696546 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036021.1 NP_001031098.1 4 - 5 685 96
NM_001036022.1 NP_001031099.1 4 - 5 856 96
NM_102549.5 NP_174105.2 4 - 5 1038 96
GenBank mRNA AK175747.1 - 4 - 5 677 96
AK175917.1 - 4 - 5 677 96
AK176142.1 - 4 - 5 677 96
AY057556.1 - 4 - 5 856 96
AY074265.1 - 4 - 5 856 96
AY096624.1 - 4 - 5 749 96

View Full Product Details