Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PUB45
Entrez Gene ID
839684
Gene Name
plant U-box 45
Gene Aliases
ARABIDOPSIS THALIANA PLANT U-BOX 45, AT1G27910, ATPUB45, F13K9.2, F13K9_2, plant U-box 45
Chromosome Location
Chr.1: 9720733 - 9724800 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102556.5 NP_174112.1 5 - 6 1935 146
GenBank mRNA AY094469.1 - 5 - 6 1847 146
BT002721.1 - 4 - 5 1503 146

View Full Product Details