Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HB-1
Entrez Gene ID
839740
Gene Name
homeobox-1
Gene Aliases
AT1G28420, F3M18.14, F3M18_14, RINGLET 1, RLT1, homeobox-1
Chromosome Location
Chr.1: 9979478 - 9987737 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102610.4 NP_174164.2 3 - 4 1188 81

View Full Product Details