Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SFP2
Entrez Gene ID
832795
Gene Name
Major facilitator superfamily protein
Gene Aliases
AT5G27360, F21A20.70, F21A20_70
Chromosome Location
Chr.5: 9656949 - 9662663 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344004.1 NP_001330510.1 15 - 16 1501 110
NM_001344005.1 NP_001330511.1 16 - 17 1348 110
NM_001344007.1 NP_001330509.1 13 - 14 1278 110
NM_122618.3 NP_568494.1 16 - 17 1298 110
GenBank mRNA AY026255.1 - 16 - 17 1195 110

View Full Product Details