Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALY1
Entrez Gene ID
832823
Gene Name
protein ALWAYS EARLY 1
Gene Aliases
ALWAYS EARLY 1, ARABIDOPSIS THALIANA ALWAYS EARLY 1, AT5G27610, ATALY1, F15A18.70, F15A18_70
Chromosome Location
Chr.5: 9764135 - 9769992 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122643.3 NP_198113.2 8 - 9 1219 143
GenBank mRNA AB493764.1 - 7 - 8 988 143

View Full Product Details