Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYCH;1
Entrez Gene ID
832824
Gene Name
cyclin H;1
Gene Aliases
AT5G27620, F15A18.80, F15A18_80, cyclin H;1
Chromosome Location
Chr.5: 9771287 - 9774920 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344019.1 NP_001330469.1 4 - 5 759 128
NM_001344020.1 NP_001330468.1 4 - 5 759 128
NM_122644.5 NP_198114.2 4 - 5 775 128
GenBank mRNA AB051072.1 - 4 - 5 300 128
AK118707.1 - 4 - 5 757 128

View Full Product Details