Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EGY1
Entrez Gene ID
833476
Gene Name
Peptidase M50 family protein
Gene Aliases
AMOS1, AT5G35220, ETHYLENE-DEPENDENT GRAVITROPISM-DEFICIENT AND YELLOW-GREEN 1, T25C13.100, T25C13_100, ammonium overly sensitive 1
Chromosome Location
Chr.5: 13484403 - 13487594 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122913.4 NP_198372.1 3 - 4 988 127
GenBank mRNA AY050809.1 - 3 - 4 941 127
BT001978.1 - 3 - 4 940 127

View Full Product Details