Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCR80
Entrez Gene ID
833815
Gene Name
low-molecular-weight cysteine-rich 80
Gene Aliases
AT5G38330, MSI17.50, MSI17_50, low-molecular-weight cysteine-rich 80
Chromosome Location
Chr.5: 15317683 - 15318511 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123194.3 NP_198649.2 1 - 2 144 86
GenBank mRNA DQ447011.1 - 1 - 2 60 86

View Full Product Details