The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_123283.5 | NP_568563.1 | 2 - 3 | 350 | 72 |
GenBank mRNA | AY065367.1 | - | 2 - 3 | 208 | 72 |
AY114028.1 | - | 2 - 3 | 197 | 72 |