Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RABG1
Entrez Gene ID
833958
Gene Name
RAB GTPase homolog G1
Gene Aliases
AT5G39620, AtRABG1, MIJ24.90, MIJ24_90, RAB GTPase homolog G1
Chromosome Location
Chr.5: 15864166 - 15865782 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123324.1 NP_568566.1 5 - 6 403 105
GenBank mRNA AB071853.1 - 5 - 6 403 105

View Full Product Details