Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WNK8
Entrez Gene ID
834204
Gene Name
with no lysine (K) kinase 8
Gene Aliases
AT5G41990, ATWNK8, EIP1, EMF1-Interacting Protein 1, MJC20.9, MJC20_9, with no lysine (K) kinase 8
Chromosome Location
Chr.5: 16794967 - 16798006 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123564.3 NP_568599.1 1 - 2 536 92
GenBank mRNA AF424629.1 - 1 - 2 536 92
AY113042.1 - 1 - 2 92 92

View Full Product Details