Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl41
Entrez Gene ID
834228
Gene Name
serine carboxypeptidase-like 41
Gene Aliases
AT5G42230, K5J14.3, K5J14_3, serine carboxypeptidase-like 41
Chromosome Location
Chr.5: 16880592 - 16884462 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344440.1 NP_001318730.1 8 - 9 1235 113
NM_123588.4 NP_199038.1 7 - 8 1395 113
GenBank mRNA BT002948.1 - 7 - 8 1028 113
BT004386.1 - 6 - 7 897 113

View Full Product Details