Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ckl7
Entrez Gene ID
834433
Gene Name
casein kinase I-like 7
Gene Aliases
AT5G44100, MLN1.2, MLN1_2, casein kinase I-like 7
Chromosome Location
Chr.5: 17749057 - 17752849 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344533.1 NP_001330873.1 12 - 13 1353 61
NM_123777.4 NP_199223.1 11 - 12 1477 61
GenBank mRNA AY943846.1 - 11 - 12 923 61
BT015795.1 - 11 - 12 923 61
BT021097.1 - 11 - 12 1019 61

View Full Product Details