Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAO1
Entrez Gene ID
834478
Gene Name
target of AVRB operation1
Gene Aliases
AT5G44510, MFC16.19, MFC16_19, target of AVRB operation1
Chromosome Location
Chr.5: 17929558 - 17934222 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344552.1 NP_001330713.1 2 - 3 1650 112
NM_001344553.1 NP_001330714.1 2 - 3 1650 112
NM_001344554.1 NP_001330712.1 2 - 3 1650 112
NM_123818.2 NP_199264.1 2 - 3 1684 112

View Full Product Details