Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RTL3
Entrez Gene ID
834551
Gene Name
RNAse THREE-like protein 3
Gene Aliases
AT5G45150, K18C1.3, K18C1_3, RNAse THREE-like protein 3
Chromosome Location
Chr.5: 18260313 - 18264454 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344592.1 NP_001330579.1 6 - 7 2810 79
NM_001344593.1 NP_001330578.1 6 - 7 2735 79
NM_123883.2 NP_199328.1 7 - 8 2572 79

View Full Product Details