Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FKBP13
Entrez Gene ID
834607
Gene Name
FK506-binding protein 13
Gene Aliases
AT5G45680, ATFKBP13, FK506 BINDING PROTEIN 13, FK506-binding protein 13, MRA19.7, MRA19_7
Chromosome Location
Chr.5: 18530834 - 18532337 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123935.5 NP_199380.1 4 - 5 620 69
GenBank mRNA AY065047.1 - 4 - 5 577 69
AY091680.1 - 4 - 5 560 69

View Full Product Details