Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DOG1
Entrez Gene ID
834623
Gene Name
delay of germination 1
Gene Aliases
AT5G45830, ATDOG1, DELAY OF GERMINATION 1, GAAS5, GLUCOSE SENSING QTL 5, GSQ5, K15I22.3, K15I22_3, germination ability after storage 5
Chromosome Location
Chr.5: 18589418 - 18591506 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344641.1 NP_001318745.1 2 - 3 903 67
NM_123951.3 NP_199395.2 2 - 3 871 67

View Full Product Details