Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- KCO3
- Entrez Gene ID
-
834679
- Gene Name
- Ca2+ activated outward rectifying K+ channel 3
- Gene Aliases
-
AT5G46360, ATKCO3, CA2+ ACTIVATED OUTWARD RECTIFYING K+ CHANNEL 3, Ca2+ activated outward rectifying K+ channel 3, MPL12.16, MPL12_16
- Chromosome Location
-
Chr.5: 18806563 - 18808224 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124006.3 |
NP_199448.1 |
1 - 2 |
720 |
131 |
GenBank mRNA |
AJ010873.1 |
- |
1 - 2 |
720 |
131 |
View Full Product Details