Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RPS6
- Entrez Gene ID
-
834690
- Gene Name
- disease resistance protein (TIR-NBS-LRR class) family
- Gene Aliases
-
AT5G46470, K11I1.6, K11I1_6, RESISTANT TO P. SYRINGAE 6
- Chromosome Location
-
Chr.5: 18842553 - 18849741 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124017.3 |
NP_199459.2 |
4 - 5 |
2414 |
78 |
View Full Product Details