Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LON2
- Entrez Gene ID
-
834750
- Gene Name
- lon protease 2
- Gene Aliases
-
ABERRANT PEROXISOME MORPHOLOGY 10, APEM10, AT5G47040, MQD22.18, MQD22_18, lon protease 2
- Chromosome Location
-
Chr.5: 19092858 - 19098772 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124075.4 |
NP_568675.1 |
9 - 10 |
1274 |
68 |
View Full Product Details