Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBOHD
Entrez Gene ID
834842
Gene Name
respiratory burst oxidase homologue D
Gene Aliases
AT5G47910, ATRBOHD, MCA23.25, MCA23_25, RESPIRATORY BURST OXIDASE, respiratory burst oxidase homologue D
Chromosome Location
Chr.5: 19397041 - 19402061 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124165.3 NP_199602.1 1 - 2 1169 69
GenBank mRNA AF424625.1 - 1 - 2 767 69
BT002651.1 - 1 - 2 625 69

View Full Product Details