Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- THAS1
- Entrez Gene ID
-
834852
- Gene Name
- thalianol synthase 1
- Gene Aliases
-
AT5G48010, Arabidopsis thaliana thalianol synthase 1, AtTHAS1, MDN11.9, MDN11_9, THALIANOL SYNTHASE, THAS, thalianol synthase 1
- Chromosome Location
-
Chr.5: 19456837 - 19461706 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124175.4 |
NP_199612.3 |
11 - 12 |
1687 |
83 |
GenBank mRNA |
AY327541.1 |
- |
11 - 12 |
1646 |
83 |
View Full Product Details