Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THAS1
Entrez Gene ID
834852
Gene Name
thalianol synthase 1
Gene Aliases
AT5G48010, Arabidopsis thaliana thalianol synthase 1, AtTHAS1, MDN11.9, MDN11_9, THALIANOL SYNTHASE, THAS, thalianol synthase 1
Chromosome Location
Chr.5: 19456837 - 19461706 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085264.2 NP_001078733.1 1 - 2 366 79
NM_124175.4 NP_199612.3 1 - 2 243 79
GenBank mRNA AY327541.1 - 1 - 2 202 79

View Full Product Details