Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sks3
Entrez Gene ID
834900
Gene Name
SKU5 similar 3
Gene Aliases
AT5G48450, MJE7.8, MJE7_8, SKU5 similar 3, SKU5 similar 3
Chromosome Location
Chr.5: 19632703 - 19635679 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344773.1 NP_001332438.1 6 - 7 1433 99
NM_124220.3 NP_199656.2 6 - 7 1418 99

View Full Product Details