Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OVA2
Entrez Gene ID
834962
Gene Name
tRNA synthetase class I (I, L, M and V) family protein
Gene Aliases
AT5G49030, K19E20.18, K19E20_18, ovule abortion 2
Chromosome Location
Chr.5: 19874925 - 19883297 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036965.2 NP_001032042.1 5 - 6 777 92
NM_001203568.1 NP_001190497.1 6 - 7 1210 92
NM_124280.4 NP_199714.2 5 - 6 889 92
GenBank mRNA AY080830.1 - 5 - 6 706 92
AY142605.1 - 5 - 6 606 92

View Full Product Details