Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MET1
Entrez Gene ID
834975
Gene Name
methyltransferase 1
Gene Aliases
AT5G49160, DDM2, DECREASED DNA METHYLATION 2, DMT01, DMT1, DNA METHYLTRANSFERASE, DNA METHYLTRANSFERASE 01, DNA METHYLTRANSFERASE 1, MET2, METHYLTRANSFERASE 2, METHYLTRANSFERASE I, METI, methyltransferase 1
Chromosome Location
Chr.5: 19932114 - 19938470 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344823.1 NP_001331134.1 6 - 7 3198 59
NM_124293.4 NP_199727.1 5 - 6 3507 59
GenBank mRNA L10692.1 - 5 - 6 3400 59

View Full Product Details